Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226862
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
5
613
NM_001321198.1
NP_001308127.1
5
494
NM_001321199.1
NP_001308128.1
5
458
NM_001321200.1
NP_001308129.1
5
514
NM_001321201.1
NP_001308130.1
4
386
NM_018224.3
NP_060694.2
4
393
NR_047701.1
5
458
NR_135580.1
5
494
NR_135581.1
5
494
NR_135582.1
4
393
NR_135583.1
4
393
XM_005249807.4
XP_005249864.1
4
457
XM_006715752.2
XP_006715815.1
5
425
XM_017012413.1
XP_016867902.1
4
384
XM_017012414.1
XP_016867903.1
4
384
XM_017012415.1
XP_016867904.1
5
486
XM_017012416.1
XP_016867905.1
4
384
XM_017012417.1
XP_016867906.1
4
384
XM_017012418.1
XP_016867907.1
4
384
XM_017012419.1
XP_016867908.1
4
384
XM_017012420.1
XP_016867909.1
5
486
XM_017012421.1
XP_016867910.1
5
608
XM_017012422.1
XP_016867911.1
5
497
XM_017012423.1
XP_016867912.1
5
417
XM_017012424.1
XP_016867913.1
4
400
XM_017012425.1
XP_016867914.1
5
501
XM_017012426.1
XP_016867915.1
5
620
XM_017012427.1
XP_016867916.1
6
575
XM_017012428.1
XP_016867917.1
4
400
XM_017012429.1
XP_016867918.1
3
294
XM_017012430.1
XP_016867919.1
5
486
XM_017012431.1
XP_016867920.1
4
400
XM_017012432.1
XP_016867921.1
4
400
GenBank mRNA
AK001665.1
-
5
427
AK021905.1
-
5
563
AK023007.1
-
5
410
AK023053.1
-
5
426
AK097203.1
-
3
1924
AK293043.1
-
5
436
BC056884.1
-
5
385
6159045
BG028456.1
-
415
BG334368.1
-
5
403
BG548154.1
-
5
450
BM918741.1
-
4
366
BP377872.1
-
4
393
BQ231567.1
-
4
331
DA742258.1
-
4
375
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0