Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 3 | 287 | ||
NR_111943.1 | 2 | 493 | ||||
NR_111944.1 | 3 | 287 | ||||
GenBank mRNA | AK311951.1 | - | 3 | 195 | ||
BC009407.2 | - | 1 | 178 | 4334552 | ||
BC019899.1 | - | 1 | 183 | 3882924 | ||
BC020453.1 | - | 1 | 182 | |||
BC022370.1 | - | 3 | 200 | 4738331 | ||
BC049824.1 | - | 1 | 175 | 6212543 | ||
BC062715.1 | - | 3 | 285 | 4068842 | ||
BC070222.1 | - | 1 | 196 | 6738383 | ||
BC071928.1 | - | 1 | 186 | 5090053 | ||
BE905890.1 | - | 2 | 163 | |||
BI597659.1 | - | 2 | 493 | |||
BM017772.1 | - | 3 | 288 | |||
BM457623.1 | - | 2 | 180 | |||
DA310472.1 | - | 2 | 493 | |||
M13932.1 | - | 3 | 196 |