Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 17 | 3855 | ||
NM_001164815.1 | NP_001158287.1 | 16 | 3774 | |||
NM_001164816.1 | NP_001158288.1 | 10 | 1413 | |||
NM_001164817.1 | NP_001158289.1 | 11 | 1475 | |||
NM_014977.3 | NP_055792.1 | 17 | 3894 | |||
XM_005267415.3 | XP_005267472.1 | 17 | 3695 | |||
XM_005267416.3 | XP_005267473.1 | 17 | 3662 | |||
XM_005267418.1 | XP_005267475.1 | 10 | 1710 | |||
GenBank mRNA | AB014570.2 | - | 17 | 3403 | ||
AF124726.1 | - | 17 | 3893 | |||
AF124727.1 | - | 11 | 1429 | |||
AF124728.1 | - | 10 | 1404 | |||
AK299034.1 | - | 16 | 3285 | |||
AL050382.1 | - | 10 | 2499 | |||
AL833228.1 | - | 12 | 1744 | |||
BC140805.1 | - | 17 | 3624 | 9021673 | ||
BX247975.1 | - | 10 | 1683 |