Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIMCH1 SIRNA Assays ›
Gene Symbol:
LIMCH1
Gene Name:
LIM and calponin homology domains 1
Gene Aliases:
LIMCH1A, LMO7B
Chromosome Location:
Chr.4: 41359607 - 41700044 on Build GRCh38
UniGene ID:
Hs.335163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22806
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIMCH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112717.2
NP_001106188.1
12
1420
NM_001112718.2
NP_001106189.1
12
1420
NM_001112719.2
NP_001106190.1
7
1205
NM_001112720.2
NP_001106191.1
6
1169
NM_001289122.1
NP_001276051.1
11
1384
NM_001289124.1
NP_001276053.1
11
1429
NM_014988.3
NP_055803.2
12
1420
XM_005248057.1
XP_005248114.1
19
3052
XM_005248058.1
XP_005248115.1
19
3052
XM_005248060.1
XP_005248117.1
19
3052
XM_005248061.1
XP_005248118.1
19
3052
XM_005248067.2
XP_005248124.1
17
3926
XM_005248072.1
XP_005248129.1
12
1420
XM_005248074.1
XP_005248131.1
11
1384
XM_005248075.1
XP_005248132.1
11
1384
XM_006713996.1
XP_006714059.1
19
3052
XM_011513642.1
XP_011511944.1
19
3052
XM_011513643.1
XP_011511945.1
19
3052
XM_011513644.1
XP_011511946.1
19
3052
XM_011513645.1
XP_011511947.1
19
3052
XM_011513646.1
XP_011511948.1
19
3052
XM_011513648.1
XP_011511950.1
18
2809
XM_011513649.1
XP_011511951.1
14
2711
XM_011513651.2
XP_011511953.1
17
3931
XM_011513653.1
XP_011511955.1
12
1420
XM_011513654.1
XP_011511956.1
11
1384
XM_011513655.1
XP_011511957.1
6
1043
XM_011513656.1
XP_011511958.1
7
1079
XM_011513657.1
XP_011511959.1
6
1043
XM_011513658.2
XP_011511960.1
11
1645
XM_017007894.1
XP_016863383.1
19
3052
XM_017007895.1
XP_016863384.1
19
3052
XM_017007896.1
XP_016863385.1
19
3052
XM_017007897.1
XP_016863386.1
19
3052
XM_017007898.1
XP_016863387.1
12
1420
XM_017007899.1
XP_016863388.1
12
1420
XM_017007900.1
XP_016863389.1
12
1420
XM_017007901.1
XP_016863390.1
12
1420
XM_017007902.1
XP_016863391.1
12
1420
XM_017007903.1
XP_016863392.1
7
1079
XM_017007904.1
XP_016863393.1
7
1079
XM_017007905.1
XP_016863394.1
7
1079
GenBank mRNA
AB029025.2
-
12
1418
AK294774.1
-
6
1232
AK295784.1
-
5
758
AK295836.1
-
6
868
AK298915.1
-
7
1075
AK299297.1
-
6
1039
AK299835.1
-
6
903
AK309846.1
-
6
1111
AL831962.2
-
17
2807
BC023546.2
-
9
1273
4110496
BX537916.1
-
6
1039
BX640692.1
-
5
797
CR749205.1
-
7
1075
CR933645.1
-
12
1421
CR936601.1
-
11
1355
CR936610.1
-
11
1430
CR936658.1
-
12
1399
CR936661.1
-
11
1324
CR936664.1
-
12
1421
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-rzq2w:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0