Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIMCH1 SIRNA Assays ›
Gene Symbol:
LIMCH1
Gene Name:
LIM and calponin homology domains 1
Gene Aliases:
LIMCH1A, LMO7B
Chromosome Location:
Chr.4: 41359607 - 41700044 on Build GRCh38
UniGene ID:
Hs.335163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22807
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIMCH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112717.2
NP_001106188.1
25
3154
NM_001112718.2
NP_001106189.1
25
3151
NM_001112719.2
NP_001106190.1
20
2936
NM_001112720.2
NP_001106191.1
19
2900
NM_001289122.1
NP_001276051.1
25
3190
NM_001289124.1
NP_001276053.1
25
3235
NM_014988.3
NP_055803.2
26
3232
XM_005248057.1
XP_005248114.1
33
4873
XM_005248058.1
XP_005248115.1
33
4870
XM_005248060.1
XP_005248117.1
33
4861
XM_005248061.1
XP_005248118.1
32
4789
XM_005248067.2
XP_005248124.1
31
5735
XM_005248072.1
XP_005248129.1
26
3229
XM_005248074.1
XP_005248131.1
25
3193
XM_005248075.1
XP_005248132.1
24
3115
XM_006713996.1
XP_006714059.1
32
4783
XM_011513642.1
XP_011511944.1
34
4948
XM_011513643.1
XP_011511945.1
34
4945
XM_011513644.1
XP_011511946.1
34
4939
XM_011513645.1
XP_011511947.1
33
4864
XM_011513646.1
XP_011511948.1
33
4861
XM_011513648.1
XP_011511950.1
33
4705
XM_011513649.1
XP_011511951.1
29
4607
XM_011513651.2
XP_011511953.1
32
5827
XM_011513653.1
XP_011511955.1
27
3316
XM_011513654.1
XP_011511956.1
26
3280
XM_011513655.1
XP_011511957.1
21
2939
XM_011513656.1
XP_011511958.1
21
2897
XM_011513657.1
XP_011511959.1
20
2852
XM_011513658.2
XP_011511960.1
24
3376
XM_017007894.1
XP_016863383.1
34
4936
XM_017007895.1
XP_016863384.1
33
4864
XM_017007896.1
XP_016863385.1
32
4777
XM_017007897.1
XP_016863386.1
31
4702
XM_017007898.1
XP_016863387.1
26
3241
XM_017007899.1
XP_016863388.1
26
3238
XM_017007900.1
XP_016863389.1
25
3157
XM_017007901.1
XP_016863390.1
25
3148
XM_017007902.1
XP_016863391.1
24
3070
XM_017007903.1
XP_016863392.1
22
2975
XM_017007904.1
XP_016863393.1
21
2888
XM_017007905.1
XP_016863394.1
20
2813
GenBank mRNA
AB029025.2
-
26
3230
AK027231.1
-
12
1156
AK294774.1
-
20
3041
AK295784.1
-
19
2567
AK295836.1
-
20
2677
AK299297.1
-
19
2770
AK299835.1
-
20
2712
AK302674.1
-
12
1402
AK309846.1
-
19
2842
AL831962.2
-
31
4616
BC023546.2
-
22
3004
4110496
BC029735.1
-
10
906
BC095394.1
-
15
1451
BX537916.1
-
19
2770
BX640692.1
-
18
2528
CR749205.1
-
20
2806
CR936601.1
-
25
3161
CR936610.1
-
25
3236
CR936658.1
-
22
2902
CR936661.1
-
24
3055
CR936664.1
-
25
3152
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-zl9nd:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0