Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS1 SIRNA Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
UniGene ID:
Hs.485729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22810
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168408.1
NP_001161880.1
11, 12
1515
NM_001168409.1
NP_001161881.1
11, 12
1300
NM_001168410.1
NP_001161882.1
11, 12
1587
NM_014989.5
NP_055804.2
16, 17
3102
XM_005248688.3
XP_005248745.1
16, 17
3038
XM_017010516.1
XP_016866005.1
16, 17
3038
XM_017010517.1
XP_016866006.1
16, 17
3038
XM_017010519.1
XP_016866008.1
16, 17
3038
XM_017010520.1
XP_016866009.1
16, 17
3038
XM_017010521.1
XP_016866010.1
16, 17
3038
XM_017010522.1
XP_016866011.1
16, 17
3038
XM_017010523.1
XP_016866012.1
16, 17
3038
XM_017010525.1
XP_016866014.1
16, 17
3038
XM_017010526.1
XP_016866015.1
16, 17
3038
XM_017010529.1
XP_016866018.1
16, 17
3038
XM_017010530.1
XP_016866019.1
16, 17
3038
XM_017010532.1
XP_016866021.1
16, 17
3038
XM_017010533.1
XP_016866022.1
16, 17
3038
XM_017010534.1
XP_016866023.1
11, 12
1611
XM_017010536.1
XP_016866025.1
11, 12
1608
XM_017010537.1
XP_016866026.1
11, 12
1608
XM_017010538.1
XP_016866027.1
11, 12
1716
XM_017010539.1
XP_016866028.1
11, 12
1603
XM_017010540.1
XP_016866029.1
11, 12
1602
XM_017010542.1
XP_016866031.1
11, 12
1602
XM_017010546.1
XP_016866035.1
11, 12
1714
XM_017010548.1
XP_016866037.1
11, 12
1599
XM_017010550.1
XP_016866039.1
11, 12
1982
XM_017010552.1
XP_016866041.1
11, 12
1839
GenBank mRNA
AB002338.1
-
17, 18
2825
AB045726.1
-
17, 18
3068
AB051866.1
-
17, 18
3068
AF263305.1
-
4, 5
456
AF263306.1
-
4, 5
456
AF263308.1
-
4, 5
456
AK293828.1
-
11, 12
1190
AK295001.1
-
11, 12
1433
AK296303.1
-
11, 12
1356
AK302571.1
-
1, 2
53
AK308658.1
-
11, 12
1587
AK309185.1
-
11, 12
1300
AK316108.1
-
11, 12
1313
AY190519.1
-
16, 17
2781
BC151853.1
-
17, 18
2849
8860502
BC152435.1
-
17, 18
2825
8862556
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0