Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYT1L SIRNA Assays ›
Gene Symbol:
MYT1L
Gene Name:
myelin transcription factor 1 like
Gene Aliases:
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location:
Chr.2: 1789113 - 2331388 on Build GRCh38
UniGene ID:
Hs.434418
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22898
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYT1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303052.1
NP_001289981.1
10
1666
NM_015025.3
NP_055840.2
10
1666
XM_011510318.1
XP_011508620.1
10
1728
XM_011510319.1
XP_011508621.1
10
1838
XM_011510320.2
XP_011508622.1
10
1769
XM_011510321.2
XP_011508623.1
10
1719
XM_011510322.2
XP_011508624.1
10
1532
XM_011510323.1
XP_011508625.1
9
1451
XM_011510324.1
XP_011508626.1
10
1728
XM_011510325.1
XP_011508627.1
10
1728
XM_011510326.1
XP_011508628.1
10
1728
XM_011510327.1
XP_011508629.1
10
1728
XM_011510328.2
XP_011508630.1
10
1728
XM_011510331.2
XP_011508633.1
10
1728
XM_011510332.2
XP_011508634.1
10
1728
XM_017003604.1
XP_016859093.1
11
1817
XM_017003605.1
XP_016859094.1
10
1396
XM_017003606.1
XP_016859095.1
10
1838
XM_017003607.1
XP_016859096.1
10
1769
XM_017003608.1
XP_016859097.1
10
1728
XM_017003609.1
XP_016859098.1
10
1713
XM_017003610.1
XP_016859099.1
11
1812
XM_017003611.1
XP_016859100.1
10
1728
XM_017003612.1
XP_016859101.1
10
1838
XM_017003613.1
XP_016859102.1
10
1838
XM_017003614.1
XP_016859103.1
10
1677
XM_017003615.1
XP_016859104.1
10
1728
XM_017003616.1
XP_016859105.1
10
1728
XM_017003617.1
XP_016859106.1
10
1728
XM_017003618.1
XP_016859107.1
10
1728
XM_017003619.1
XP_016859108.1
10
1838
XM_017003620.1
XP_016859109.1
3
718
XM_017003621.1
XP_016859110.1
10
1728
XM_017003622.1
XP_016859111.1
10
1728
GenBank mRNA
AB029029.2
-
11
1451
AF036943.1
-
10
1561
AK299571.1
-
6
883
BC031690.1
-
2
415
BC137272.1
-
10
1469
9021269
BC137273.1
-
10
1469
9021270
BC150281.1
-
11
1451
9007267
BF530661.1
-
415
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nervous system development
cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0