Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015092.4 | NP_055907.3 | 36 | 5962 | ||
XM_005255182.3 | XP_005255239.1 | 36 | 5975 | |||
XM_005255183.3 | XP_005255240.1 | 37 | 6033 | |||
XM_005255184.3 | XP_005255241.1 | 35 | 5819 | |||
XM_011545769.2 | XP_011544071.1 | 36 | 5975 | |||
XM_011545770.1 | XP_011544072.1 | 24 | 3772 | |||
XM_017023066.1 | XP_016878555.1 | 36 | 5785 | |||
XM_017023067.1 | XP_016878556.1 | 24 | 3835 | |||
GenBank mRNA | AB007881.2 | - | 4 | 580 | ||
AB061371.1 | - | 37 | 5914 | |||
AF186377.1 | - | 11 | 1943 | |||
AF395444.1 | - | 34 | 5269 | |||
AK074359.1 | - | 3 | 441 | |||
AY014957.1 | - | 24 | 3841 |