Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099681.2 | NP_001093151.2 | 3 | 273 | ||
NM_001099682.2 | NP_001093152.2 | 3 | 273 | |||
NM_024859.3 | NP_079135.3 | 3 | 273 | |||
XM_005278062.4 | XP_005278119.1 | 3 | 357 | |||
XM_005278063.4 | XP_005278120.1 | 3 | 356 | |||
XM_005278064.4 | XP_005278121.1 | 2 | 133 | |||
XM_005278065.3 | XP_005278122.1 | 2 | 221 | |||
XM_006724560.3 | XP_006724623.1 | 3 | 357 | |||
XM_011543985.2 | XP_011542287.1 | 3 | 354 | |||
XM_011543986.2 | XP_011542288.1 | 2 | 108 | |||
XM_011543987.2 | XP_011542289.1 | 2 | 129 | |||
GenBank mRNA | AK025340.1 | - | 2 | 55 | ||
AK094251.1 | - | 1 | 805 | |||
AK096058.1 | - | 2 | 1295 | |||
BC027995.1 | - | 3 | 242 | |||
DA072017.1 | - | 2 | 55 |