Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015043.3 | NP_055858.2 | 14 | 2395 | ||
NM_198868.2 | NP_942568.2 | 14 | 2395 | |||
GenBank mRNA | AB014576.1 | - | 14 | 2395 | ||
AB449893.1 | - | 14 | 2358 | |||
AK074791.1 | - | 4 | 477 | |||
BC008919.2 | - | 3 | 377 | |||
BC080659.1 | - | 3 | 456 | |||
BC110400.1 | - | 5 | 615 | |||
BC129994.1 | - | 14 | 2399 | 40129194 |