Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181643.5 | NP_857594.2 | 3 | 290 | ||
XM_005270472.1 | XP_005270529.1 | 3 | 399 | |||
GenBank mRNA | BC029237.1 | - | 2 | 179 | ||
BC050319.1 | - | 3 | 254 | 4837146 | ||
BC068603.1 | - | 3 | 214 | |||
BC093891.1 | - | 3 | 277 | 7939736 | ||
BC101501.1 | - | 3 | 278 | 8069007 | ||
BF674731.1 | - | 3 | 292 | |||
HM237139.1 | - | 3 | 277 | |||
HM237140.1 | - | 3 | 203 |