Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 3 | 299 | ||
GenBank mRNA | AK130965.1 | - | 3 | 288 | ||
AK312123.1 | - | 3 | 288 | |||
BC014459.1 | - | 1 | 279 | 4872948 | ||
BC058041.1 | - | 3 | 291 | 6716166 | ||
BG476195.1 | - | 3 | 408 | |||
BP382007.1 | - | 3 | 299 | |||
L13799.1 | - | 2 | 249 | |||
U02032.1 | - | 3 | 277 | |||
U37230.1 | - | 1 | 298 | |||
U43701.1 | - | 3 | 296 |