Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100620.1 | NP_001094090.1 | 2 | 292 | ||
NM_001278324.1 | NP_001265253.1 | 2 | 292 | |||
NM_005480.3 | NP_005471.3 | 2 | 292 | |||
XM_006719181.1 | XP_006719244.1 | 2 | 236 | |||
XM_011537723.1 | XP_011536025.1 | 2 | 236 | |||
XM_011537724.1 | XP_011536026.1 | 2 | 225 | |||
XM_011537725.2 | XP_011536027.1 | 3 | 428 | |||
GenBank mRNA | AK296811.1 | - | 2 | 225 | ||
AK299302.1 | - | 2 | 197 | |||
AK309716.1 | - | 2 | 225 | |||
AK310709.1 | - | 2 | 223 | |||
AK315211.1 | - | 2 | 236 | |||
BC011597.2 | - | 2 | 160 | |||
BC032583.1 | - | 2 | 204 | 5498800 | ||
DC410015.1 | - | 2 | 292 | |||
U04810.2 | - | 2 | 235 |