Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 6 | 1025 | ||
NM_001111113.1 | NP_001104583.1 | 5 | 1001 | |||
NM_001312646.1 | NP_001299575.1 | 5 | 971 | |||
NM_001312647.1 | NP_001299576.1 | 7 | 1066 | |||
NM_001312648.1 | NP_001299577.1 | 6 | 1060 | |||
NM_005339.4 | NP_005330.1 | 7 | 1154 | |||
GenBank mRNA | AB022435.1 | - | 7 | 956 | ||
AB022436.1 | - | 5 | 803 | |||
AK093533.1 | - | 7 | 975 | |||
AK291454.1 | - | 5 | 906 | |||
AK295846.1 | - | 5 | 906 | |||
AK307130.1 | - | 6 | 965 | |||
U58522.1 | - | 7 | 873 |