Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113324.2 | NP_001106795.2 | 4 | 861 | ||
GenBank mRNA | BC047782.1 | - | 4 | 858 | 6500495 | |
BX648576.1 | - | 4 | 827 |
Gene Symbol: TEN1-CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037709.1 | 4 | 861 | |||
GenBank mRNA | AK097104.1 | - | 4 | 818 | ||
BX647274.1 | - | 4 | 847 |