Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017561.1 | NP_060031.1 | 7 | 1876 | ||
GenBank mRNA | AL133071.1 | - | 6 | 1429 | ||
BC130390.1 | - | 7 | 1876 | 40146421 | ||
BC143798.1 | - | 7 | 1876 | 9052312 |
Gene Symbol: NUTM2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170741.1 | NP_001164212.1 | 7 | 1831 |