Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136478.3 | NP_001129950.1 | 3 | 694 | ||
NM_001136479.2 | NP_001129951.1 | 4 | 493 | |||
NM_001280559.1 | NP_001267488.1 | 4 | 817 | |||
NM_030920.4 | NP_112182.1 | 4 | 817 | |||
XM_005245513.3 | XP_005245570.1 | 4 | 815 | |||
XM_005245514.3 | XP_005245571.1 | 4 | 815 | |||
XM_017002418.1 | XP_016857907.1 | 4 | 815 | |||
GenBank mRNA | AK092672.1 | - | 4 | 778 | ||
AK095228.1 | - | 4 | 493 | |||
AK223469.1 | - | 4 | 684 | |||
AK303392.1 | - | 3 | 560 | |||
AL832664.1 | - | 4 | 683 | |||
AL832674.1 | - | 4 | 819 | |||
AL833660.1 | - | 4 | 571 | |||
AY057381.1 | - | 4 | 756 | |||
BC003380.1 | - | 4 | 636 | 3047915 |