Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 4 | 965 | ||
NM_001286126.1 | NP_001273055.1 | 4 | 965 | |||
NM_001286128.1 | NP_001273057.1 | 4 | 965 | |||
NM_018650.4 | NP_061120.3 | 4 | 965 | |||
XM_005273134.4 | XP_005273191.1 | 4 | 832 | |||
XM_006711326.3 | XP_006711389.1 | 4 | 226 | |||
XM_011509561.2 | XP_011507863.1 | 4 | 961 | |||
XM_011509562.2 | XP_011507864.1 | 4 | 578 | |||
XM_017001305.1 | XP_016856794.1 | 5 | 422 | |||
XM_017001306.1 | XP_016856795.1 | 4 | 462 | |||
XM_017001307.1 | XP_016856796.1 | 5 | 862 | |||
GenBank mRNA | AB040910.1 | - | 4 | 922 | ||
AF154845.1 | - | 4 | 591 | |||
AK295506.1 | - | 4 | 463 | |||
AK314614.1 | - | 4 | 625 | |||
BC113869.1 | - | 4 | 501 | 40036731 | ||
BC114478.1 | - | 4 | 501 | 40036730 | ||
KJ534879.1 | - | 4 | 325 | |||
KJ534881.1 | - | 4 | 325 | |||
KJ535089.1 | - | 4 | 324 |