Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320892.1 | NP_001307821.1 | 2 | 165 | ||
NM_022552.4 | NP_072046.2 | 2 | 165 | |||
NM_175629.2 | NP_783328.1 | 2 | 236 | |||
NM_175630.1 | NP_783329.1 | 2 | 236 | |||
NR_135490.1 | 2 | 236 | ||||
XM_005264175.4 | XP_005264232.1 | 2 | 99 | |||
XM_011532663.2 | XP_011530965.1 | 2 | 175 | |||
XM_011532664.2 | XP_011530966.1 | 2 | 176 | |||
XM_017003526.1 | XP_016859015.1 | 2 | 207 | |||
GenBank mRNA | AB208833.1 | - | 2 | 135 | ||
AF067972.2 | - | 2 | 127 | |||
AF331856.1 | - | 2 | 115 | |||
AL543971.3 | - | 2 | 165 | |||
BC032392.1 | - | 2 | 236 | |||
BC043617.1 | - | 2 | 135 | 6150112 | ||
BC051864.1 | - | 2 | 104 |