Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011828.3 | NP_035958.3 | 4 | 859 | ||
GenBank mRNA | AF060178.1 | - | 4 | 537 | ||
AF169243.1 | - | 4 | 526 | |||
AK088698.1 | - | 4 | 835 | |||
AK165112.1 | - | 4 | 825 | |||
BC025443.1 | - | 4 | 787 | 5068428 | ||
BC059008.1 | - | 4 | 616 | 6849136 |