Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081376.1 | NP_001074845.1 | 18 | 3051 | ||
NM_029216.2 | NP_083492.2 | 18 | 3051 | |||
GenBank mRNA | AK147563.1 | - | 18 | 3052 |