Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297592.1 | NP_001284521.1 | 5 | 1060 | ||
NM_001297594.1 | NP_001284523.1 | 5 | 1082 | |||
NM_015187.4 | NP_056002.2 | 5 | 1277 | |||
XM_011513819.2 | XP_011512121.2 | 5 | 1271 | |||
XM_017007923.1 | XP_016863412.1 | 4 | 915 | |||
GenBank mRNA | AB018289.1 | - | 4 | 775 | ||
AK289687.1 | - | 5 | 1025 | |||
BC060867.1 | - | 5 | 1070 | 30344157 | ||
CR749817.1 | - | 5 | 1060 |