Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008955.1 | NP_032981.1 | 3, 4 | 637 | ||
GenBank mRNA | AF017453.1 | - | 3, 4 | 637 | ||
AK076186.1 | - | 3, 4 | 646 | |||
BC099477.1 | - | 3, 4 | 633 | 30790014 | ||
DQ058643.1 | - | 3, 4 | 637 |