Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013802.3 | NP_001013824.2 | 2 | 329 | ||
NM_028387.1 | NP_082663.1 | 2 | 324 | |||
GenBank mRNA | AK004072.1 | - | 2 | 281 | ||
AK011824.1 | - | 2 | 319 | |||
AK013491.1 | - | 2 | 326 | |||
AK029184.1 | - | 2 | 304 | |||
AK134694.1 | - | 2 | 331 | |||
AK160433.1 | - | 2 | 304 | |||
AK165589.1 | - | 2 | 319 | |||
BC027552.1 | - | 2 | 164 | |||
BC028443.1 | - | 2 | 164 | |||
BC038317.1 | - | 2 | 250 |