Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028848.3 | NP_083124.1 | 1 | 245 | ||
NM_029067.1 | NP_083343.1 | 1 | 245 | |||
GenBank mRNA | AK050925.1 | - | 1 | 246 | ||
AY747687.1 | - | 1 | 220 | |||
BC089520.1 | - | 1 | 220 | 6774024 |