Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105063.1 | NP_001098533.1 | 3 | 1175 | ||
GenBank mRNA | AF011415.1 | - | 788 |
Gene Symbol: Vmn2r49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105156.1 | NP_001098626.1 | 3 | 1175 |
Set Membership: |
G-protein coupled receptor |