Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134752.1 | NP_001128224.1 | 4 | 579 | ||
GenBank mRNA | AK046418.1 | - | 4 | 580 |