Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm11554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099313.2 | NP_001092783.1 | 1 | 624 | ||
GenBank mRNA | BC104358.1 | - | 1 | 629 | 40049658 | |
BC104359.1 | - | 1 | 629 | 40049667 | ||
BC119472.1 | - | 1 | 629 | 40049663 | ||
BC119473.1 | - | 1 | 629 | 40049668 |
Gene Symbol: Gm11569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099312.1 | NP_001092782.1 | 1 | 594 |
Gene Symbol: Gm11595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126322.1 | NP_001119794.1 | 1 | 921 |
Gene Symbol: Gm11596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099311.2 | NP_001092781.1 | 1 | 608 |
Gene Symbol: Krtap4-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027087.3 | NP_081363.1 | 1 | 553 | ||
GenBank mRNA | AK009035.1 | - | 1 | 554 |
Gene Symbol: Krtap4-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026834.2 | NP_081110.1 | 1 | 639 | ||
GenBank mRNA | AK004258.1 | - | 1 | 640 | ||
BC120646.1 | - | 1 | 606 | |||
BC120648.1 | - | 1 | 606 |
Gene Symbol: Krtap4-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085548.2 | NP_001079017.1 | 1 | 780 |