Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CREM SIRNA Assays ›
Gene Symbol:
Crem
Gene Name:
cAMP responsive element modulator
Gene Aliases:
Icer
Chromosome Location:
Chr.17: 57021704 - 57090888 on Build Rnor_6.0
UniGene ID:
Rn.10251
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s234985
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Crem
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001110860.2
NP_001104330.1
10
1757
NM_001271101.1
NP_001258030.1
10
1361
NM_001271102.1
NP_001258031.1
3
711
NM_001271245.1
NP_001258174.1
4
397
NM_001271246.1
NP_001258175.1
4
855
NM_001271247.1
NP_001258176.1
4
901
NM_001271248.1
NP_001258177.1
3
423
NM_017334.2
NP_059030.2
4
351
XM_006254049.2
XP_006254111.1
9
1715
XM_006254050.2
XP_006254112.1
9
1319
XM_006254051.2
XP_006254113.1
9
1541
XM_006254052.2
XP_006254114.1
8
1873
XM_006254053.2
XP_006254115.1
9
1495
XM_006254054.2
XP_006254116.1
8
1595
XM_006254055.2
XP_006254117.1
8
1459
XM_006254056.2
XP_006254118.1
8
1063
XM_006254057.2
XP_006254119.1
9
1562
XM_006254058.2
XP_006254120.1
8
1526
XM_006254059.2
XP_006254121.1
8
1130
XM_006254060.2
XP_006254122.1
7
1726
XM_006254061.2
XP_006254123.1
7
1330
XM_006254062.2
XP_006254124.1
6
1294
XM_006254063.2
XP_006254125.1
7
1445
XM_006254064.2
XP_006254126.1
8
1306
XM_006254065.2
XP_006254127.1
8
910
XM_006254066.2
XP_006254128.1
7
1270
XM_006254067.2
XP_006254129.1
7
874
XM_006254068.2
XP_006254130.1
5
1105
XM_006254069.2
XP_006254131.1
4
1134
XM_006254070.2
XP_006254132.1
3
1223
XM_008771691.1
XP_008769913.1
4
1180
XM_008771692.1
XP_008769914.1
4
776
XM_008771693.1
XP_008769915.1
3
1418
XM_008771694.1
XP_008769916.1
3
755
XM_008771695.1
XP_008769917.1
3
734
GenBank mRNA
BC078899.1
-
10
1361
7133070
CO556486.1
-
3
372
FQ213544.1
-
3
711
FQ219343.1
-
3
708
FQ220921.1
-
3
710
S66024.1
-
4
348
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001110859, NM_001110856, NM_013498, NM_001110853, NM_001271505, and others.
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
spermatogenesis
cAMP-mediated signaling
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0