Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047885.1 | NP_001041350.1 | 3 | 217 | ||
XM_006232280.2 | XP_006232342.1 | 4 | 404 | |||
XM_006232281.2 | XP_006232343.1 | 4 | 409 | |||
GenBank mRNA | BC168644.1 | - | 3 | 519 |