Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 9 | 1189 | ||
NM_015264.1 | NP_056079.1 | 9 | 1261 | |||
XM_011530017.2 | XP_011528319.1 | 9 | 1224 | |||
XM_017028672.1 | XP_016884161.1 | 9 | 1233 | |||
GenBank mRNA | AB023147.1 | - | 8 | 957 | ||
AK025608.1 | - | 7 | 778 | |||
AK094359.1 | - | 1 | 142 | |||
AK094937.1 | - | 10 | 1272 | |||
AK124648.1 | - | 9 | 1269 | |||
AK297058.1 | - | 9 | 1241 | |||
AK309015.1 | - | 8 | 1313 | |||
AK315883.1 | - | 9 | 1173 | |||
AY007105.1 | - | 2 | 143 | |||
BC016882.1 | - | 4 | 519 | |||
BC038414.1 | - | 8 | 1336 | |||
BC053595.1 | - | 9 | 1183 | 5730020 | ||
CR456402.1 | - | 9 | 1126 |