Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 9, 10 | 1233 | ||
NM_015264.1 | NP_056079.1 | 9, 10 | 1305 | |||
XM_011530017.2 | XP_011528319.1 | 9, 10 | 1268 | |||
XM_017028672.1 | XP_016884161.1 | 9, 10 | 1277 | |||
GenBank mRNA | AB023147.1 | - | 8, 9 | 1001 | ||
AK025608.1 | - | 7, 8 | 822 | |||
AK094937.1 | - | 10, 11 | 1316 | |||
AK124648.1 | - | 9, 10 | 1313 | |||
AK297058.1 | - | 9, 10 | 1285 | |||
AK309015.1 | - | 8, 9 | 1357 | |||
AK315883.1 | - | 9, 10 | 1217 | |||
AY007105.1 | - | 2, 3 | 187 | |||
BC016882.1 | - | 4, 5 | 563 | |||
BC038414.1 | - | 8, 9 | 1380 | |||
BC053595.1 | - | 9, 10 | 1227 | 5730020 | ||
CR456402.1 | - | 9, 10 | 1170 |