Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108071.1 | NP_001101541.1 | 3 | 330 | ||
XM_006240915.2 | XP_006240977.1 | 5 | 1259 | |||
XM_006240916.2 | XP_006240978.1 | 5 | 1249 | |||
XM_006240917.2 | XP_006240979.1 | 5 | 1256 | |||
XM_006240918.2 | XP_006240980.1 | 5 | 1199 | |||
XM_006240919.2 | XP_006240981.1 | 5 | 1258 | |||
XM_006240920.2 | XP_006240982.1 | 4 | 1088 | |||
GenBank mRNA | FQ224249.1 | - | 5 | 559 |