Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ost4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134690.1 | NP_001128162.1 | 2 | 405 | ||
NM_001134691.1 | NP_001128163.1 | 3 | 305 | |||
GenBank mRNA | BC157814.1 | - | 2 | 383 | 8362186 | |
CB706964.1 | - | 3 | 266 |