Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134983.1 | NP_001128455.1 | 3 | 1394 | ||
XM_006235424.2 | XP_006235486.1 | 2 | 1211 | |||
XM_006235425.2 | XP_006235487.1 | 3 | 3283 | |||
XM_006235426.2 | XP_006235488.1 | 3 | 1458 | |||
XM_006235427.2 | XP_006235489.1 | 4 | 1363 | |||
GenBank mRNA | BC168719.1 | - | 3 | 1394 | 8364226 | |
FQ235168.1 | - | 3 | 1332 |