Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320867.1 | NP_001307796.1 | 5 | 624 | ||
NM_001320868.1 | NP_001307797.1 | 4 | 508 | |||
NM_001320869.1 | NP_001307798.1 | 3 | 363 | |||
NM_014297.4 | NP_055112.2 | 5 | 657 | |||
XM_005258687.3 | XP_005258744.1 | 5 | 754 | |||
GenBank mRNA | AK309880.1 | - | 5 | 602 | ||
AK315346.1 | - | 5 | 620 | |||
BC008250.1 | - | 5 | 607 | 3872059 | ||
BG706505.1 | - | 4 | 457 | |||
D83198.2 | - | 5 | 646 | |||
KU178565.1 | - | 5 | 578 | |||
KU178566.1 | - | 5 | 545 | |||
KU178567.1 | - | 3 | 284 |