Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018957.3 | NP_061830.3 | 16, 17 | 1701 | ||
GenBank mRNA | AK124370.1 | - | 17, 18 | 2594 | ||
AK126556.1 | - | 15, 16 | 2396 | |||
AL157480.1 | - | 5, 6 | 525 | |||
AY364241.1 | - | 13, 14 | 1401 | |||
BC008282.1 | - | 14, 15 | 1494 | 3639782 | ||
CR456576.1 | - | 16, 17 | 1620 |