Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105203.1 | NP_001098673.1 | 10 | 2787 | ||
NM_001105204.1 | NP_001098674.1 | 10 | 2469 | |||
NM_001105205.1 | NP_001098675.1 | 9 | 1809 | |||
NM_001278227.1 | NP_001265156.1 | 9 | 1753 | |||
NM_001278228.1 | NP_001265157.1 | 8 | 1989 | |||
NM_001278229.1 | NP_001265158.1 | 9 | 1534 | |||
NM_001278230.1 | NP_001265159.1 | 8 | 1651 | |||
NM_014328.4 | NP_055143.2 | 9 | 1852 | |||
NR_103478.1 | 8 | 1199 | ||||
XM_006711254.1 | XP_006711317.1 | 11 | 3198 | |||
XM_006711256.1 | XP_006711319.1 | 11 | 2880 | |||
XM_006711257.1 | XP_006711320.1 | 9 | 1466 | |||
XM_017000891.1 | XP_016856380.1 | 10 | 3368 | |||
XM_017000892.1 | XP_016856381.1 | 9 | 1148 | |||
GenBank mRNA | AB026894.2 | - | 9 | 1276 | ||
AK055451.1 | - | 9 | 2047 | |||
AK074485.1 | - | 8 | 1087 | |||
AK074982.1 | - | 9 | 1271 | |||
AK093777.1 | - | 7 | 2111 | |||
AK125378.1 | - | 10 | 2421 | |||
AK298730.1 | - | 9 | 1263 | |||
AK307736.1 | - | 8 | 1438 | |||
AK314559.1 | - | 9 | 1521 | |||
BC001045.2 | - | 9 | 1293 | |||
BC004293.1 | - | 8 | 1055 | |||
BC028055.1 | - | 8 | 1401 | |||
BC052277.1 | - | 9 | 1809 | |||
BX402179.2 | - | 9 | 1023 | |||
BX640612.1 | - | 8 | 1469 | |||
CR749671.1 | - | 8 | 2358 |