Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUSC1 SIRNA Assays ›
Gene Symbol:
RUSC1
Gene Name:
RUN and SH3 domain containing 1
Gene Aliases:
NESCA
Chromosome Location:
Chr.1: 155320849 - 155331118 on Build GRCh38
UniGene ID:
Hs.226499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24222
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUSC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105203.1
NP_001098673.1
9, 10
2763
NM_001105204.1
NP_001098674.1
9, 10
2445
NM_001105205.1
NP_001098675.1
8, 9
1785
NM_001278227.1
NP_001265156.1
8, 9
1729
NM_001278228.1
NP_001265157.1
7, 8
1965
NM_001278229.1
NP_001265158.1
8, 9
1510
NM_001278230.1
NP_001265159.1
7, 8
1627
NM_014328.4
NP_055143.2
8, 9
1828
NR_103478.1
7, 8
1175
XM_006711254.1
XP_006711317.1
10, 11
3174
XM_006711256.1
XP_006711319.1
10, 11
2856
XM_006711257.1
XP_006711320.1
8, 9
1442
XM_017000891.1
XP_016856380.1
9, 10
3344
XM_017000892.1
XP_016856381.1
8, 9
1124
GenBank mRNA
AB026894.2
-
8, 9
1252
AK055451.1
-
8, 9
2023
AK074485.1
-
7, 8
1063
AK074982.1
-
8, 9
1247
AK093777.1
-
6, 7
2087
AK125378.1
-
9, 10
2397
AK298730.1
-
8, 9
1239
AK307736.1
-
7, 8
1414
AK314559.1
-
8, 9
1497
AL080083.1
-
7, 8
1105
BC001045.2
-
8, 9
1269
BC004293.1
-
7, 8
1031
BC028055.1
-
7, 8
1377
BC052277.1
-
8, 9
1785
BX402179.2
-
8, 9
999
BX640612.1
-
7, 8
1445
CR749671.1
-
7, 8
2334
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
positive regulation of signal transduction
actin binding
SH3/SH2 adaptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0