Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 12 | 1002 | ||
NM_001256733.1 | NP_001243662.1 | 11 | 918 | |||
NM_001256734.1 | NP_001243663.1 | 11 | 912 | |||
NM_001256735.1 | NP_001243664.1 | 11 | 888 | |||
NM_001256736.1 | NP_001243665.1 | 11 | 888 | |||
NM_012446.3 | NP_036578.2 | 12 | 978 | |||
XM_011543297.2 | XP_011541599.1 | 12 | 917 | |||
XM_017009304.1 | XP_016864793.1 | 12 | 820 | |||
XM_017009305.1 | XP_016864794.1 | 13 | 909 | |||
XM_017009306.1 | XP_016864795.1 | 12 | 893 | |||
XM_017009307.1 | XP_016864796.1 | 11 | 706 | |||
XM_017009308.1 | XP_016864797.1 | 13 | 1081 | |||
XM_017009309.1 | XP_016864798.1 | 12 | 1145 | |||
XM_017009310.1 | XP_016864799.1 | 12 | 1293 | |||
GenBank mRNA | AF077048.1 | - | 12 | 842 | ||
AF161465.1 | - | 12 | 808 | |||
AK074537.1 | - | 12 | 773 | |||
AK293544.1 | - | 11 | 713 | |||
AK297494.1 | - | 12 | 686 | |||
AK299858.1 | - | 11 | 756 | |||
AK312337.1 | - | 12 | 822 | |||
AL080076.1 | - | 12 | 826 | |||
BC017020.1 | - | 12 | 826 | 3913337 | ||
CR936787.1 | - | 11 | 733 | |||
DA202987.1 | - | 4 | 168 |