Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282739.1 | NP_001269668.1 | 1 | 737 | ||
NM_030758.3 | NP_110385.1 | 1 | 737 | |||
XM_005261465.4 | XP_005261522.1 | 1 | 663 | |||
XM_006724203.2 | XP_006724266.2 | 1 | 663 | |||
XM_011530057.1 | XP_011528359.1 | 1 | 663 | |||
XM_011530058.1 | XP_011528360.1 | 1 | 663 | |||
XM_011530059.2 | XP_011528361.1 | 1 | 663 | |||
XM_017028708.1 | XP_016884197.1 | 1 | 663 | |||
XM_017028709.1 | XP_016884198.1 | 1 | 663 | |||
XM_017028711.1 | XP_016884200.1 | 1 | 633 | |||
XM_017028713.1 | XP_016884202.1 | 1 | 459 | |||
GenBank mRNA | AB051451.2 | - | 1 | 645 | ||
AF288741.1 | - | 1 | 631 | |||
AF323731.1 | - | 1 | 737 | |||
AK097581.1 | - | 1 | 669 | |||
AK294007.1 | - | 1 | 669 | |||
BC118914.2 | - | 1 | 654 | 40037933 |