Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017526.1 | NP_001017526.1 | 4 | 426 | ||
NM_001198726.1 | NP_001185655.1 | 4 | 426 | |||
NM_181335.2 | NP_851852.2 | 4 | 426 | |||
GenBank mRNA | AF544240.1 | - | 3 | 285 | ||
AK001306.1 | - | 4 | 426 | |||
AK022305.1 | - | 3 | 368 | |||
BC010490.2 | - | 2 | 270 | |||
BC048280.1 | - | 4 | 378 | 5190702 | ||
BC059382.1 | - | 4 | 410 |
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181334.5 | NP_851851.3 | 7 | 955 | ||
GenBank mRNA | AF195968.1 | - | 6 | 546 | ||
AF195969.1 | - | 5 | 461 | |||
BC132755.1 | - | 5 | 446 | 40146777 | ||
BC132757.1 | - | 5 | 446 | 40146779 | ||
BC144382.1 | - | 5 | 446 | 9052903 |