Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012420.2 | NP_036552.1 | 2 | 1324 | ||
GenBank mRNA | AK293346.1 | - | 3 | 986 | ||
AK312358.1 | - | 2 | 1157 | |||
BC025786.1 | - | 2 | 1303 | 5229306 | ||
CR457031.1 | - | 1 | 1093 | |||
U34605.1 | - | 2 | 1210 |