Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167621.1 | NP_001161093.1 | 5, 6 | 810 | ||
NM_013236.3 | NP_037368.1 | 6, 7 | 1002 | |||
GenBank mRNA | AF086923.1 | - | 6, 7 | 945 | ||
AF119662.1 | - | 6, 7 | 942 | |||
AK025734.1 | - | 6, 7 | 933 | |||
AK095309.1 | - | 6, 7 | 770 | |||
AK294348.1 | - | 5, 6 | 803 | |||
AL050282.1 | - | 6, 7 | 934 | |||
BC007508.2 | - | 6, 7 | 924 | 3030062 | ||
CR456568.1 | - | 6, 7 | 787 | |||
CR457381.1 | - | 6, 7 | 736 |