Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015393.3 | NP_056208.2 | 2 | 392 | ||
XM_011531833.1 | XP_011530135.1 | 3 | 456 | |||
GenBank mRNA | AK022311.1 | - | 2 | 351 | ||
AL080121.1 | - | 2 | 309 | |||
AY358777.1 | - | 2 | 337 | |||
BC013294.2 | - | 2 | 321 | 3028327 | ||
CA390477.1 | - | 2 | 389 |