Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001991.2 | NP_001001991.1 | 6 | 1216 | ||
NM_001282675.1 | NP_001269604.1 | 8 | 1206 | |||
NM_001282676.1 | NP_001269605.1 | 4 | 883 | |||
NM_015430.3 | NP_056245.2 | 6 | 1216 | |||
GenBank mRNA | AF370388.1 | - | 5 | 704 | ||
AK297092.1 | - | 8 | 1207 | |||
AK300185.1 | - | 4 | 468 | |||
AK316188.1 | - | 4 | 455 | |||
AL832391.1 | - | 6 | 782 | |||
AY358346.1 | - | 6 | 795 | |||
BC089434.1 | - | 6 | 808 | 30558617 | ||
BX640676.1 | - | 6 | 1216 |