Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFD1L SIRNA Assays ›
Gene Symbol:
MTHFD1L
Gene Name:
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Gene Aliases:
FTHFSDC1, MTC1THFS, dJ292B18.2
Chromosome Location:
Chr.6: 150865679 - 151101887 on Build GRCh38
UniGene ID:
Hs.591343
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24712
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFD1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242767.1
NP_001229696.1
15
1717
NM_001242768.1
NP_001229697.1
15
1401
NM_015440.4
NP_056255.2
15
1714
XM_005266907.4
XP_005266964.1
15
1655
XM_005266910.4
XP_005266967.1
15
1658
XM_005266911.4
XP_005266968.1
15
1658
XM_011535729.2
XP_011534031.1
15
1658
XM_011535730.2
XP_011534032.1
15
1401
XM_011535731.2
XP_011534033.1
15
1373
XM_011535732.2
XP_011534034.1
15
1455
XM_011535733.2
XP_011534035.1
15
1901
XM_011535734.2
XP_011534036.1
15
1494
XM_011535737.2
XP_011534039.1
15
1658
XM_011535738.2
XP_011534040.1
15
1658
XM_017010702.1
XP_016866191.1
15
1657
XM_017010703.1
XP_016866192.1
15
1657
XM_017010704.1
XP_016866193.1
15
1373
XM_017010705.1
XP_016866194.1
15
1898
XM_017010706.1
XP_016866195.1
15
1901
XM_017010707.1
XP_016866196.1
15
1898
XM_017010708.1
XP_016866197.1
15
1659
GenBank mRNA
AB127387.1
-
15
1570
AK127089.1
-
14
2193
AK315378.1
-
14
1510
AL117452.1
-
15
1389
AY374130.1
-
15
1677
BC017477.1
-
10
961
BC110319.1
-
14
1336
BC136685.1
-
15
1607
9020675
BC144368.1
-
15
1607
9052889
BC144369.1
-
15
1604
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
one-carbon metabolic process
folic acid-containing compound metabolic process
folic acid-containing compound biosynthetic process
formate metabolic process
tetrahydrofolate interconversion
tetrahydrofolate metabolic process
folic acid metabolic process
embryonic neurocranium morphogenesis
embryonic viscerocranium morphogenesis
oxidation-reduction process
formate-tetrahydrofolate ligase activity
methenyltetrahydrofolate cyclohydrolase activity
methylenetetrahydrofolate dehydrogenase (NADP+) activity
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0