Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136191.2 | NP_001129663.1 | 7, 8 | 2129 | ||
NM_015493.6 | NP_056308.3 | 5, 6 | 1871 | |||
GenBank mRNA | AB040951.2 | - | 6, 7 | 2081 | ||
AB284125.1 | - | 8, 9 | 2312 | |||
AK002094.1 | - | 4, 5 | 561 | |||
AK023332.1 | - | 4, 5 | 613 | |||
AY639929.1 | - | 5, 6 | 1792 | |||
BC082762.1 | - | 7, 8 | 2083 | 6497026 | ||
BC098105.1 | - | 6, 7 | 1881 | 40012030 | ||
BC098286.1 | - | 6, 7 | 1881 | 40012033 | ||
BC098312.1 | - | 6, 7 | 1881 | 40012032 |