Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 9 | 1257 | ||
NM_032107.4 | NP_115479.4 | 12 | 1428 | |||
GenBank mRNA | AB014581.1 | - | 5 | 507 | ||
AK299199.1 | - | 12 | 1398 | |||
AL110279.1 | - | 1 | 1949 | |||
BC039820.1 | - | 9 | 1172 | 5199689 |