Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015650.3 | NP_056465.2 | 9 | 1295 | ||
XM_011510944.2 | XP_011509246.1 | 10 | 1452 | |||
XM_011510946.2 | XP_011509248.1 | 9 | 1380 | |||
XM_011510947.2 | XP_011509249.1 | 10 | 1439 | |||
XM_011510948.2 | XP_011509250.1 | 8 | 1254 | |||
XM_011510950.2 | XP_011509252.1 | 2 | 255 | |||
XM_017003789.1 | XP_016859278.1 | 10 | 1452 | |||
GenBank mRNA | AF242456.1 | - | 9 | 1394 |